Tisk…
Pipette canisters, flat bottom
Dodavatel: BELLCO GLASS
1290-00250 1290-38250 1290-38430 1290-00300
BELC1290-00250EA 7480 CZK
BELC1290-00250 BELC1290-38250 BELC1290-38430 BELC1290-00300
Pipette canisters, flat bottom
Nádoby Válce na pipety
Glass pipette canisters constructed of borosilicate glass.
  • With flat bottom
  • All components will withstand repeated dry or steam heat sterilization
  • Bottom silicone pad prevents chipping of pipette tips