Tisk…
SP Bel-Art Glove box, portable
Dodavatel: Bel-Art Products, a Part of SP
H500282000 H500282001
BELAH500282000EA 102780 CZK
BELAH500282000 BELAH500282001
SP Bel-Art Glove box, portable
Gloveboxy
This lightweight, self-contained portable isolation cabinet made of LDPE protects the operator and the external environment.
  • Large acrylic window with 6" circular glove ports permits safe viewing of operations
  • 8” diameter side port allows easy insertion of material and equipment
  • Large size, 22" long (20" installed) neoprene gloves (included for some models only) provide good protection against gas, liquid or solid penetration
  • Gloves are secured to the 6" circular port holes with clamping rings (included for some models only)
  • Body molded from LDPE with
More Product Information