Tisk…
Sterilising trays and covers, SP Bel-Art
Dodavatel: Bel-Art Products, a Part of SP
H162640000 H162630000 H162620000 H162600000 H162610000
BELAH162640000EA 3110 CZK
BELAH162640000 BELAH162630000 BELAH162620000 BELAH162600000 BELAH162610000
Sterilising trays and covers, SP Bel-Art
Podnosy/Misky/Kelímky Podnosy sterilizační
These lightweight and corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
  • Ideal for sterilising instruments, bottles containing agar, deionised water, saline and other reagents, and for safely cleaning glassware in sinks
  • Polypropylene covers are 3,2 mm thick with a 25,4 mm skirt that fits snugly over the rim of the tray
  • Trays and lids are steam autoclavable at 121 ºC