Tisk…
Pipette tray, SP Bel-Art
Dodavatel: Bel-Art Products, a Part of SP
F189400000
BELAF189400000EA 2350 CZK
BELAF189400000
Pipette tray, SP Bel-Art
Podnosy/Misky/Kelímky Podnosy na pipety
This pipette tray made of PS can be used in drawers or on the benchtop for storing pipettes and can also be used for soaking pipettes before cleaning.
  • Scalloped rims hold 16 small or 7 large pipettes
  • Center support holds 12 short pipettes in a perpendicular direction