Tisk…
Wash bottles, narrow or wide neck, with cap, SP Bel-Art
Dodavatel: Bel-Art Products, a Part of SP
F116510016 F116500008 F116500016
BELAF116510016EA 2460 CZK
BELAF116510016 BELAF116500008 BELAF116500016
Wash bottles, narrow or wide neck, with cap, SP Bel-Art
Láhve Střičky
LDPE, red with red PP cap.
Bottles and caps are vividly coloured to aid in the identification of hazardous contents.