Tisk…
Kjeldhal block digestion systems
Dodavatel: BEHR
B00605456 B00632822
BEHRB00605456EA 146760 CZK
BEHRB00605456 BEHRB00632822
Kjeldhal block digestion systems
Systémy rozkladové Mineralizátory
Block digestion systems with high quality corrosion-resistant block housing made of stainless steel. With 8, 12 or 20 sample slots for standard Kjeldahl digestion vessels with a volume of 250 ml, has high efficiency heating and extraction hood with exhaust collector.
  • Behr single-knob control for particularly easy and fast programming
  • Menu navigation in national language.
  • 10 freely configurable programs for block temperature and digestion time
  • Applications can be saved
  • USB interface available
  • The Windows software enables bi-directional transfer application-
More Product Information