Tisk…
Deionisers, behropur®
Dodavatel: BEHR
B00223462 B00223463
BEHRB00223462EA 18970 CZK
BEHRB00223462 BEHRB00223463
Deionisers, behropur®
Systémy pro výrobu čisté vody Deionizátroy
These deionisers are thick-walled, robust and practical mixed-bed ion exchangers made of blue polyethylene with free output to the storage tank. Ideal also for subsequent demineralisation of reverse osmosis system or for an ambitious aquarist.
  • Can be connected directly to the water supply and is immediately ready for use
  • Automatic deaeration due to the water being supplied from the bottom
  • Nozzles are sturdy and easy to clean
  • Minimum risk of microbial contamination due to slit nozzles in the raw water supply
  • Extremely secure and leakproof sealing of the
More Product Information
Objednat
Produktová specifikace
Model B5 B10
Environmentally Preferable
Kapacita 500 L 1000 L
Maximální průtok 50 l/h 100 l/h
Ø×H 160×530 mm 210×630 mm