Tisk…
Plasma collection tube with heparin, BD Vacutainer®
Dodavatel: BD Medical
368480 368886 367526 368884 367869 368495 367876
BDAM368480EA 2650 CZK
BDAM368480 BDAM368886 BDAM367526 BDAM368884 BDAM367869 BDAM368495 BDAM367876
Plasma collection tube with heparin, BD Vacutainer®
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET or glass, clear. Lithium Heparin/Sodium Heparin tubes.
  • Centrifugation conditions: ≤1300xg for 10 minutes at 18 to 25 °C
  • Transparent or paper label
BD Vacutainer® plasma tubes for clinical chemistry are available with spray-dried sodium heparin or lithium heparin additives. Heparin acts as an anticoagulant as it develops an antithrombin complex.
The optimum anticoagulation is achieved in all BD Vacutainer® tubes by the use of 17 IU pharmaceutical grade heparin per mL of blood when the fill level is correct. The lithium heparin in BD Vacutainer®More Product Information