Tisk…
3-Piece Syringes
Dodavatel: BD Medical
300188 305502 308062 303179
613-3913EA 0 CZK
613-3913 BDAM305502 613-1253 BDAM303179
3-Piece Syringes
Stříkačky Střílkačky zdravotnické
BD Plastipak™ syringe is a precision drug delivery device that is an essential part of your daily patient care toolkit, which you can use across a range of critical applications and departments. It is intended for general purpose injection and aspiration of fluids from vials, ampoules and parts of the body below the surface of the skin.
  • 3-piece design with latex-free stopper
  • Clear barrel for visualization of syringe content
  • Bold scale printing to aid in setting for a more accurate dosage of medication
  • Special silicone lubrication ensures that plunger moves
More Product Information