Tisk…
Centrifuge tubes, Falcon®
Dodavatel: Corning
352196
BDAA352196EA 3780 CZK
BDAA352196
Centrifuge tubes, Falcon®
Zkumavky Zkumavky centrifugační
PP, high clarity, conical, printed graduation, with dome seal PE screw caps, sterile.
  • Dome seal screw cap offer both vented and fully closed positions
  • RNase/DNase-free
Withstands forces up to 12000×g.