Tisk…

SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)

Dodavatel: G Bioscienes
Varování

Synonyms: SMPB

BC21
BC21EA 17380 CZK
BC21
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)

Succinimidyl 4-(ρ-maleimidophenyl) butyrate Heterobifunctional Cross Linkers.


  • Ideal for enzyme-antibody crosslinking

Vzorec: C₁₈H₁₆N₂O₆
MW: 356,33 g/mol
MDL Number: MFCD00054961
CAS číslo: 79886-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dc2f348b167b5c3af824f83c0eec294e49051c4&vid=0dc2f348b167b5c3af824f83c0eec294e49051c4&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Spacer Arm (Å) 11.6
Reactive Toward Primary Amine + Sulhydryl
Membrane Permeable X
Water Soluble No
Cleavable/ Reversible No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR