Tisk…

Sulpho-GMBS (N-Maleimidobutyryloxysulphosuccinimide ester)

Dodavatel: G Bioscienes

Synonyms: Sulfo GMBS

BC14
BC14EA 28870 CZK
BC14
Sulpho-GMBS (N-Maleimidobutyryloxysulphosuccinimide ester)
Sulpho-GMBS (N-Maleimidobutyryloxysulphosuccinimide ester)

N-γ-Maleimidobutyryloxysulfosuccinimide ester Heterobifunctional Cross Linker.

Vzorec: C₁₂H₁₂N₂O₉S
MW: 360.297 g/mol
MDL Number: MFCD00153605
CAS číslo: 158018-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a47a06ed71fbc1f39b64a02fe2105bbde695e13d&vid=a47a06ed71fbc1f39b64a02fe2105bbde695e13d&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Spacer Arm (Å) 6.8
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable No
Water Soluble Yes
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR