Tisk…
Thermo Scientific™ Locator™ Plus Rack and Box Systems
Dodavatel: Thermo Fisher Scientific
CY50925 CY509105 CY509106 CY509107 CY509108 CY509109 CY509110 CY509111 CY50925-70 CY50935 CY50935-70 CY50945-70 CY50985-70
478-0167EA 167590 CZK
478-0167 BARNCY509105 BARNCY509106 BARNCY509107 BARNCY509108 BARNCY509109 BARNCY509110 BARNCY509111 478-0265 478-0807 478-0266 478-0267 478-0268
Thermo Scientific™ Locator™ Plus Rack and Box Systems
Dewarovy a kryogenní nádoby
Locator™ cryogenic rack and box systems allow quick, efficient sample retrieval with maximum storage capacity (up to 6000 samples with Locator™ Plus) combined with minimum liquid nitrogen consumption.
  • Compact design minimises space requirements while maximising storage capacity
  • System is suited for both manual and computerised inventory record-keeping methods
  • Super-insulated vessels have special, narrow-mouth design that conserves liquid nitrogen by minimising evaporation
  • Outstanding temperature uniformity: Samples are stored below –180 °C, even when less
More Product Information