Tisk…
IntelliStack™ CO₂ Incubator Shakers
Dodavatel: Thermo Fisher Scientific
8892620D 8892620U 8895020D 8895020U
BARN8892620DEA 995720 CZK
BARN8892620D BARN8892620U BARN8895020D BARN8895020U
IntelliStack™ CO₂ Incubator Shakers
Třepačky Třepačky inkubované
The Thermo Scientific™ IntelliStack™ CO₂ Incubator shaker is a versatile laboratory instrument used for incubating and agitating samples simultaneously. It combines the functions of an incubator and shaker, providing a controlled environment for cell culture, microbial growth, and various other applications. With adjustable temperature, speed, and CO₂ concentration settings, it offers precise control over experimental conditions. A large, easy-to-use 10" colour touchscreen display with alarms, alerts, real-time monitoring of key parameters, and data logging with export via USB or Ethernet
More Product Information