Tisk…
LN₂ Supply Tanks
Dodavatel: Thermo Fisher Scientific
8120 8127
BARN8120EA 404420 CZK
BARN8120 BARN8127
LN₂ Supply Tanks
Dewarovy a kryogenní nádoby
Transport cryogenic industrial gases with Thermo Scientific™ LN₂ Supply tanks, which are designed to be both durable and user-friendly. For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilising controlled-rate freezing equipment.
  • Rated at 22 psi (151,7 kPa)
  • Evaporate 2% of volume per day
Upozornení:
Warranty varies by country, please contact us for details.