Tisk…
Ultrasonic baths, SONOREX DIGIPLUS
Dodavatel: BANDELIN-ELECTRONIC
7185 7184 7186 7181 7182 7182-GB 7184-CH 7184-GB
BAND7185EA 74450 CZK
BAND7185 BAND7184 BAND7186 BAND7181 BAND7182 BAND7182-GB BAND7184-CH BAND7184-GB
Ultrasonic baths, SONOREX DIGIPLUS
Lázně ultrazvukové
SONOREX DIGIPLUS is the high-power ultrasonic bath with fast degassing and with power settings. For degassing of HPLC solvents and for accelerating of chemical reactions, mixing of plasma and sera, for emulsifying and homogenising of samples for residue analysis in vegetarian food, preparation for pollutant analysis of either drinking or drain water, preparation of liposomes in cosmetics and pharmacy, preparation of samples for analysis of THC-content in canabis and for cleaning of technical glassware like burettes, pipettes, petri dishes and laboratory flasks and sensitive materials.