Tisk…
Ultrasonic pipette cleaners
Dodavatel: BANDELIN-ELECTRONIC
2070 2070-GB 2069-CH 2069-GB
BAND2070EA 129510 CZK
BAND2070 BAND2070-GB BAND2069-CH BAND2069-GB
Ultrasonic pipette cleaners
Lázně ultrazvukové
These ultrasonic pipette cleaners are used for gentle cleaning of glass volume-measuring devices and parts with lengths up to 755 mm. having capacity reange from 9 to 18 L.
  • Made up of stainless steel
  • Kipp handles for easy transport
  • Short cleaning times
  • Ball valve makes the emptying quickly and easily
Model PR 140 DH consist of optional heating unit have adjustable temperature range from 20 to 80 °C. This heating unit helps to increase the cleaning effect, in the case of grease residue and stubborn soiling.
Informace o dodávce: Supplied withMore Product Information