Tisk…

2-Ethoxybenzamide 97

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethenzamide

B25455.22 B25455.36
B25455.22EA 882 CZK
B25455.22 B25455.36
2-Ethoxybenzamide 97
2-Ethoxybenzamide

Purity: 97% CAS No.: 938-73-8 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 131-134° Merck Reference: 14,3731 MDL No.: MFCD00007977

Vzorec: C₉H₁₁NO₂
MW: 165.19
Bod varu: 333
Bod tání: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00007977
CAS číslo: 938-73-8
EINECS: 213-346-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR