Tisk…

Glyoxylic acid 50% (w/w) vodný roztok

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Oxoacetic acid

B25149.0B B25149.0I B25149.30
B25149.0BEA 3180 CZK
B25149.0B B25149.0I B25149.30
Glyoxylic acid 50% (w/w) vodný roztok
Glyoxylic acid

CAS No.: 298-12-4 Molecular Formula: OHCCO2H Formula Weight: 74.04 Storage Temperature: 2°C to 8°C Melting Point: -93° Boiling Point: 111° UN No.: 3265 MDL No.: MFCD00006958

Vzorec: C₂H₂O₃
MW: 74.04 g/mol
Bod varu: 111 °C
Bod tání: –93 °C
Hustota: 1.339
Storage Temperature: Refrigerator
MDL Number: MFCD00006958
CAS číslo: 298-12-4
EINECS: 206-058-5
UN: 3265
ADR: 8 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR