Tisk…

3-Fluorophenyl isothiocyanate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-Fluoro-3-isothiocyanatobenzene

B25001.06 B25001.14
B25001.06EA 1640 CZK
B25001.06 B25001.14
3-Fluorophenyl isothiocyanate ≥97%
3-Fluorophenyl isothiocyanate

Purity: 97+% CAS No.: 404-72-8 Molecular Formula: C7H4FNS Formula Weight: 153.18 Flash Point: 85°(185°F) Refractive Index: 1.6200 Odour: Pungent Boiling Point: 227° Beilstein Reference: 2081172 UN No.: 2922 MDL No.: MFCD00004804

Vzorec: C₇H₄FNS
MW: 153.18
Bod varu: 227 °C
Hustota: 1.27
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004804
CAS číslo: 404-72-8
EINECS: 206-967-7
UN: 2922
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR