Tisk…

Methyl propionylacetate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

B24848.14 B24848.22 B24848.36
B24848.14EA 1330 CZK
B24848.14 B24848.22 B24848.36
Methyl propionylacetate ≥99%
Methyl propionylacetate

Purity: 99% CAS No.: 30414-53-0 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 71°(159°F) Refractive Index: 1.4220 Boiling Point: 73-74°/5mm Beilstein Reference: 1753684 MDL No.: MFCD00011705

Vzorec: C₆H₁₀O₃
Bod varu: 73…74 °C (5 mmHg)
Hustota: 1,037
Bod vzplanutí: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011705
CAS číslo: 30414-53-0
EINECS: 250-184-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR