Tisk…

N,N-Dimethylthioformamide ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B24529.06 B24529.14 B24529.22
B24529.06EA 1250 CZK
B24529.06 B24529.14 B24529.22
N,N-Dimethylthioformamide ≥97%
N,N-Dimethylthioformamide

Purity: 97% CAS No.: 758-16-7 Molecular Formula: C3H7NS Formula Weight: 89.15 Boiling Point: 58-60°/1mm MDL No.: MFCD00004943

Vzorec: C₃H₇NS
Bod varu: 58…60 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00004943
CAS číslo: 758-16-7
EINECS: 212-060-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR