Tisk…

Ethyl (methylthio)acetate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (Methylthio)acetic ethyl ester, Ethyl 2-(methylthio)acetate

B24285.14 B24285.22
B24285.14EA 2080 CZK
B24285.14 B24285.22
Ethyl (methylthio)acetate ≥98%
Ethyl (methylthio)acetate

Purity: 98% CAS No.: 4455-13-4 Molecular Formula: CH3SCH2CO2CH2CH3 Formula Weight: 134.20 Flash Point: 59°(138°F) Refractive Index: 1.4600 Odour: Characteristic Boiling Point: 176° Beilstein Reference: 1744999 UN No.: 3272 MDL No.: MFCD00009182

Vzorec: C₅H₁₀O₂S
MW: 134.2
Bod varu: 176 °C
Hustota: 1.04
Bod vzplanutí: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009182
CAS číslo: 4455-13-4
EINECS: 224-700-2
UN: 3272
ADR: 3 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR