Tisk…

Ethyl-3-ethoxypropionate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethyl 3-ethoxypropanoate

B24024.AK B24024.AU
B24024.AKEA 906 CZK
B24024.AK B24024.AU
Ethyl-3-ethoxypropionate ≥99%
Ethyl-3-ethoxypropionate

Purity: 99+% CAS No.: 763-69-9 Molecular Formula: C7H14O3 Formula Weight: 146.19 Flash Point: 59°(142°F) Refractive Index: 1.4065 Odour: Ester-like Melting Point: -100° Boiling Point: 169-171° UN No.: 3272 MDL No.: MFCD00051356

Vzorec: C₇H₁₄O₃
MW: 146.19 g/mol
Bod varu: 169…171 °C
Bod tání: –100 °C
Hustota: 0.949
Bod vzplanutí: 59 °C (142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051356
CAS číslo: 763-69-9
EINECS: 212-112-9
UN: 3272
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR