Tisk…

Ethyl-2-hydroxyisobutyrate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B23592.14 B23592.22 B23592.36
B23592.14EA 1460 CZK
B23592.14 B23592.22 B23592.36
Ethyl-2-hydroxyisobutyrate ≥98%
Ethyl-2-hydroxyisobutyrate

Purity: 98% CAS No.: 80-55-7 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 44°(111°F) Refractive Index: 1.4080 Boiling Point: 149-150° Beilstein Reference: 878308 UN No.: 3272 MDL No.: MFCD00004458

Vzorec: C₆H₁₂O₃
Bod varu: 149…150 °C
Hustota: 0,980
Bod vzplanutí: 44 °C (111 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004458
CAS číslo: 80-55-7
EINECS: 201-290-3
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR