Tisk…

N-Ethylethylenediamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B23577.06 B23577.14
B23577.06EA 903 CZK
B23577.06 B23577.14
N-Ethylethylenediamine 98%
N-Ethylethylenediamine

Purity: 98% CAS No.: 110-72-5 Molecular Formula: CH3CH2NHCH2CH2NH2 Formula Weight: 88.15 Flash Point: 34°(93°F) Refractive Index: 1.4400 Boiling Point: 128-130° Beilstein Reference: 1098272 UN No.: 2733 MDL No.: MFCD00008166

Vzorec: C₄H₁₂N₂
Bod varu: 128…130 °C
Hustota: 0,837
Bod vzplanutí: 34 °C (93 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008166
CAS číslo: 110-72-5
EINECS: 203-795-4
UN: 2733
ADR: 3,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR