Tisk…

Aminomethanesulphonic acid 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B23441.14 B23441.22 B23441.36
B23441.14EA 766 CZK
B23441.14 B23441.22 B23441.36
Aminomethanesulphonic acid 97%
Aminomethanesulphonic acid

Purity: 97% CAS No.: 13881-91-9 Molecular Formula: H2NCH2SO3H Formula Weight: 111.12 pH: 3.2 Melting Point: ca 180° dec. Beilstein Reference: 1811756 UN No.: 2585 MDL No.: MFCD00008124

Vzorec: CH₅NO₃S
MW: 111.12
Bod tání: ∼ 180 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00008124
CAS číslo: 13881-91-9
EINECS: 237-649-6
ADR: 8 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR