Tisk…

Methyl isonicotinate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B23263.14 B23263.22 B23263.36 AB132158.0500
B23263.14EA 1050 CZK
B23263.14 B23263.22 B23263.36 ABCRAB132158.0500
Methyl isonicotinate ≥98%
Methyl isonicotinate

Purity: 98% CAS No.: 2459-09-8 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Flash Point: 82°(180°F) Refractive Index: 1.5135 Odour: Characteristic Melting Point: 8-8.5° Boiling Point: 207-209° MDL No.: MFCD00006427

Vzorec: C₇H₇NO₂
Bod varu: 207…209 °C
Hustota: 1,016
Bod vzplanutí: 82 °C (180 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006427
CAS číslo: 2459-09-8
EINECS: 219-456-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR