Tisk…

Pyromellitic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

B23099.22 B23099.36
B23099.22EA 1180 CZK
B23099.22 B23099.36
Pyromellitic acid 96%
Pyromellitic acid

Purity: 96% CAS No.: 89-05-4 Molecular Formula: C10H6O8 Formula Weight: 254.15 Melting Point: 281-285° Merck Reference: 14,8004 MDL No.: MFCD00002471

Vzorec: C₁₀H₆O₈
Bod varu: 516 °C (1013 hPa)
Bod tání: 281…285 °C
Hustota: 1,79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002471
CAS číslo: 89-05-4
EINECS: 201-879-5
Merck Index: 14,8004

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR