Tisk…

1,4-Diiodoperfluorobutane ≥97% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Varování


Stabilization: Stabilised with copper

B22660.06 B22660.14
B22660.06EA 2840 CZK
B22660.06 B22660.14
1,4-Diiodoperfluorobutane ≥97% stabilizovaný
1,4-Diiodoperfluorobutane

Stabilized with copper


Purity: 97% CAS No.: 375-50-8 Molecular Formula: C4F8I2 Formula Weight: 453.84 Refractive Index: 1.4294 Melting Point: -9° Boiling Point: 85°/100mm Beilstein Reference: 1777548 MDL No.: MFCD00042263

Vzorec: C₄F₈I₂
Bod varu: 85 °C (100 mmHg)
Bod tání: –9 °C
Hustota: 2,487
Storage Temperature: Ambient
MDL Number: MFCD00042263
CAS číslo: 375-50-8
EINECS: 206-788-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR