Tisk…

Ferulic acid methyl ester ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

B22657.04 B22657.09
B22657.04EA 2060 CZK
B22657.04 B22657.09
Ferulic acid methyl ester ≥99%
Ferulic acid methyl ester

Purity: 99% CAS No.: 2309-07-1 Molecular Formula: C11H12O4 Formula Weight: 208.22 Melting Point: 62-65° MDL No.: MFCD00017208

Vzorec: C₁₁H₁₂O₄
Bod tání: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00017208
CAS číslo: 2309-07-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR