Tisk…

2,2-Diphenylglycine ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: alpha,alpha-Diphenylglycine, Diphenylglycin, 2-Amino-2,2-diphenylacetic acid

B22650.06 B22650.14
B22650.06EA 2500 CZK
B22650.06 B22650.14
2,2-Diphenylglycine ≥98%
2,2-Diphenylglycine

Purity: 98% CAS No.: 3060-50-2 Molecular Formula: C14H13NO2 Formula Weight: 227.26 Melting Point: ca 246° dec. MDL No.: MFCD00008048

Vzorec: C₁₄H₁₃NO₂
MW: 227.26 g/mol
Bod tání: ∼ 246 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00008048
CAS číslo: 3060-50-2
EINECS: 221-299-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR