Tisk…

3,4,5-Trifluorobenzyl bromide ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 5-(Bromomethyl)-1,2,3-trifluorobenzene, α-Bromo-3,4,5-trifluorotoluene

B22578.03 B22578.06
B22578.03EA 2030 CZK
B22578.03 B22578.06
3,4,5-Trifluorobenzyl bromide ≥97%
3,4,5-Trifluorobenzyl bromide

Purity: 97% CAS No.: 220141-72-0 Molecular Formula: C7H4BrF3 Formula Weight: 225.01 Flash Point: 65°(149°F) Refractive Index: 1.5050 Boiling Point: 44-46°/1mm Beilstein Reference: 8680985 UN No.: 3265 MDL No.: MFCD00083528

Vzorec: C₇H₄BrF₃
MW: 225.01
Bod varu: 44…46 °C (1 mmHg)
Hustota: 1.689
Bod vzplanutí: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00083528
CAS číslo: 220141-72-0
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR