Tisk…

Benzyl phenyl ether ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: Benzyl Phenyl Ether

B22539.06 B22539.14 B22539.22
B22539.06EA 1670 CZK
B22539.06 B22539.14 B22539.22
Benzyl phenyl ether ≥97%
Benzyl phenyl ether

Purity: 97% CAS No.: 946-80-5 Molecular Formula: C6H5CH2OC6H5 Formula Weight: 184.24 Flash Point: 110°(230°F) Melting Point: 37-40° Boiling Point: 286-287° Beilstein Reference: 2045713 MDL No.: MFCD00020660

Vzorec: C₁₃H₁₂O
MW: 184.24 g/mol
Bod varu: 286…287 °C
Bod tání: 37…40 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020660
CAS číslo: 946-80-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR