Tisk…

Sodium-1-naphthalenesulphonate, (max. 2% H₂O) 99% (dry weight)

Dodavatel: Thermo Fisher Scientific
Varování

B22488.06 B22488.14 B22488.22
B22488.06EA 968 CZK
B22488.06 B22488.14 B22488.22
Sodium-1-naphthalenesulphonate, (max. 2% H₂O) 99% (dry weight)
Sodium-1-naphthalenesulphonate

Purity: 99% CAS No.: 130-14-3 Molecular Formula: C10H7NaO3S Formula Weight: 230.22 Melting Point: 299-301° Beilstein Reference: 3639038 MDL No.: MFCD00064964

Vzorec: C₁₀H₈O₃S
Bod tání: 299…301 °C
Storage Temperature: Ambient
MDL Number: MFCD00064964
CAS číslo: 130-14-3
EINECS: 204-976-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR