Tisk…

4-Phenoxybenzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific

B22329.03 B22329.06 B22329.14
B22329.03EA 1080 CZK
B22329.03 B22329.06 B22329.14
4-Phenoxybenzaldehyde ≥98%
4-Phenoxybenzaldehyde

Purity: 98% CAS No.: 67-36-7 Molecular Formula: C13H10O2 Formula Weight: 198.22 Flash Point: 110°(230°F) Refractive Index: 1.6110 Melting Point: 24-25° Boiling Point: 184-186°/14mm Beilstein Reference: 1947841 MDL No.: MFCD00003383

Vzorec: C₁₃H₁₀O₂
Bod varu: 184…186 °C (14 mmHg)
Bod tání: 24…25 °C
Hustota: 1,132
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003383
CAS číslo: 67-36-7
EINECS: 200-650-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR