Tisk…

trans-2-Aminocyclohexanol hydrochloride ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B22324.06 B22324.14 B22324.22
B22324.06EA 1960 CZK
B22324.06 B22324.14 B22324.22
trans-2-Aminocyclohexanol hydrochloride ≥98%
trans-2-Aminocyclohexanol hydrochloride

Purity: 98+% CAS No.: 5456-63-3 Molecular Formula: C6H13NO.HCl Formula Weight: 151.64 Melting Point: 172-175° MDL No.: MFCD00001490

Vzorec: C₆H₁₄ClNO
Bod tání: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00001490
CAS číslo: 5456-63-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR