Tisk…

Kryštálová violeť

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: CI 42555

B21932.14 B21932.22 B21932.36
B21932.14EA 872 CZK
B21932.14 B21932.22 B21932.36
Kryštálová violeť
Kryštálová violeť

CAS No.: 548-62-9 Molecular Formula: C25H30ClN3 Formula Weight: 407.99 Melting Point: ca 215° dec. Beilstein Reference: 4077708 Merck Reference: 14,4395 MDL No.: MFCD00011750

Vzorec: C₂₅H₃₀ClN₃
Hustota: 1,19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS číslo: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 14,4395

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR