Tisk…

4-Bromo-2,6-xylenol ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Bromo-2,6-dimethylphenol, 5-Bromo-2-hydroxy-3-xylene

B21669.14 B21669.22 B21669.36
B21669.14EA 1080 CZK
B21669.14 B21669.22 B21669.36
4-Bromo-2,6-xylenol ≥99%
4-Bromo-2,6-xylenol

Purity: 99% CAS No.: 2374-05-2 Molecular Formula: C8H9BrO Formula Weight: 201.07 Odour: Phenol-like Melting Point: 74-78° Beilstein Reference: 1862399 MDL No.: MFCD00002314

Vzorec: C₈H₉BrO
Bod varu: 259 °C (1013 hPa)
Bod tání: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00002314
CAS číslo: 2374-05-2
EINECS: 219-153-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR