Tisk…

(2,5-Difluorophenyl)acetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B21595.03 B21595.06 B21595.14
B21595.03EA 1040 CZK
B21595.03 B21595.06 B21595.14
(2,5-Difluorophenyl)acetic acid ≥98%
(2,5-Difluorophenyl)acetic acid

Purity: 98% CAS No.: 85068-27-5 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 123-127° Beilstein Reference: 6844956

Vzorec: (C₆H₃F₂)CH₂COOH
Bod varu: 259 °C (1013 hPa)
Bod tání: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00010000
CAS číslo: 85068-27-5
EINECS: 285-288-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR