Tisk…

10-Bromo-1-decanol 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 10-Bromodecanol

B21483.03 B21483.06
B21483.03EA 913 CZK
B21483.03 B21483.06
10-Bromo-1-decanol 95%
10-Bromo-1-decanol

Purity: 95% CAS No.: 53463-68-6 Molecular Formula: Br(CH2)10OH Formula Weight: 237.19 Flash Point: 110°(230°F) Refractive Index: 1.4760 Boiling Point: 165-169°/10mm Beilstein Reference: 1738593 MDL No.: MFCD00041681

Vzorec: HO(CH₂)₉CH₂Br
Bod varu: 165…169 °C (10 mmHg)
Hustota: 1,190
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041681
CAS číslo: 53463-68-6
EINECS: 258-572-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR