Tisk…

Tetrabutylammonium fluoride ≥75% (w/w)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B21261.14 B21261.22 B21261.36
B21261.14EA 1810 CZK
B21261.14 B21261.22 B21261.36
Tetrabutylammonium fluoride ≥75% (w/w)
Tetrabutylammonium fluoride

CAS No.: 429-41-4 Molecular Formula: [CH3(CH2)3]4NF Formula Weight: 261.47 UN No.: 2922 MDL No.: MFCD00011747

Vzorec: C₁₆H₃₆FN
Storage Temperature: Ambient
MDL Number: MFCD00011747
CAS číslo: 429-41-4
EINECS: 207-057-2
UN: 2922
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR