Tisk…

1,4-Bis(diphenylphosphino)butane ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)

B21122.36
B21122.36EA 0 CZK
B21122.36
1,4-Bis(diphenylphosphino)butane ≥98%
1,4-Bis(diphenylphosphino)butane

Purity: 98% CAS No.: 7688-25-7 Molecular Formula: C28H28P2 Formula Weight: 426.48 Melting Point: 132-136° Beilstein Reference: 706446

Vzorec: C₂₈H₂₈P₂
MW: 426.48
Bod tání: 132…136 °C
Hustota: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003051
CAS číslo: 7688-25-7
EINECS: 231-698-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR