Tisk…

Fmoc-Pro-OH 98

Dodavatel: Thermo Fisher Scientific

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

B21081.06 B21081.14
B21081.06EA 1630 CZK
B21081.06 B21081.14
Fmoc-Pro-OH 98
Fmoc-Pro-OH

Purity: 98% CAS No.: 71989-31-6 Molecular Formula: C20H19NO4 Formula Weight: 337.38 Storage Temperature: 2°C to 8°C Melting Point: 113-117° Beilstein Reference: 3596735 Optical Rotation: -40° (c=1 in ethyl acetate) MDL No.: MFCD00037122

Vzorec: C₂₀H₁₉NO₄
MW: 337.38
Bod tání: 113…117 °C
MDL Number: MFCD00037122
CAS číslo: 71989-31-6
EINECS: 276-259-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR