Tisk…

3,5-Dichlorosalicylaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

B20941.06 B20941.14 B20941.22
B20941.06EA 1400 CZK
B20941.06 B20941.14 B20941.22
3,5-Dichlorosalicylaldehyde ≥98%
3,5-Dichlorosalicylaldehyde

Purity: 98% CAS No.: 90-60-8 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 94-97° Beilstein Reference: 973391

Vzorec: C₇H₄Cl₂O₂
Bod varu: 723 °C (1013 hPa)
Bod tání: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00003320
CAS číslo: 90-60-8
EINECS: 202-005-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR