Tisk…

2-(Trifluoromethoxy)benzaldehyde 96%

Dodavatel: Thermo Fisher Scientific
Varování

B20840.06 B20840.14 B20840.22
B20840.06EA 1710 CZK
B20840.06 B20840.14 B20840.22
2-(Trifluoromethoxy)benzaldehyde 96%
2-(Trifluoromethoxy)benzaldehyde

Purity: 96% CAS No.: 94651-33-9 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Flash Point: 67°(152°F) Refractive Index: 1.4550 Boiling Point: 77°/20mm Beilstein Reference: 6137162 MDL No.: MFCD00042405

Vzorec: C₈H₅F₃O₂
MW: 190.12
Bod varu: 77 °C (20 mmHg)
Hustota: 1.330
Bod vzplanutí: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042405
CAS číslo: 94651-33-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR