Tisk…

Ethyl-3,5-dimethylisoxazole-4-carboxylate 97%

Dodavatel: Thermo Fisher Scientific

B20642.09 B20642.88
B20642.09EA 4530 CZK
B20642.09 B20642.88
Ethyl-3,5-dimethylisoxazole-4-carboxylate 97%
Ethyl-3,5-dimethylisoxazole-4-carboxylate

Purity: 97% CAS No.: 17147-42-1 Molecular Formula: C8H11NO3 Formula Weight: 169.18 Boiling Point: 87-89°/2mm MDL No.: MFCD00052557

Vzorec: C₈H₁₁NO₃
Bod varu: 87…89 °C (2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052557
CAS číslo: 17147-42-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR