Tisk…
Container with snap-on lid, Azlon®
Dodavatel: SciLabware
JWH1000P JWH2500P JWH0500P JWH0125P JWH0250P JWH2000P JWH5000P
AZLOJWH1000PEA 6880 CZK
AZLOJWH1000P AZLOJWH2500P AZLOJWH0500P AZLOJWH0125P AZLOJWH0250P AZLOJWH2000P AZLOJWH5000P
Container with snap-on lid, Azlon®
Nádobky na vzorky Nádoby pro obecné použití
PP, translucent.
  • Good chemical resistance
  • Suitable for food contact applications
  • Very good temperature resistance
These containers are lightweight tapered with tight fitting lids. Ideal for storage of samples, powders and many more.