Tisk…
Axygen® Storage boxes
Dodavatel: Corning
TR8300 TR8300-A TR8300-B TR8300-BLK
AXYGTR8300EA 2000 CZK
AXYGTR8300 AXYGTR8300-A AXYGTR8300-B AXYGTR8300-BLK
Axygen® Storage boxes
Krabice Chladící/Mrazící boxy/Sáčky
These storage boxes are the ideal solution to store samples over an extended period of time. Engineered to simplify sample identity as its polypropylene structure is meant to last.
  • Supports freezing at –80 °C and autoclaving at 121 °C
  • Alphanumeric sample identification
  • 100×1,5/2,0 ml tube capacity
These storage boxes are ideal for sample storage. These polypropylene boxes are engineered to withstand the harsh environments necessary for daily workflows in your laboratory.
The positive catch on the products anterior keeps the lid securely fastenedMore Product Information