Tisk…

Plasmid purification kit, Axygen® AxyPrep™ Mag

Dodavatel: Corning

MAG-P-S MAG-P-M
AXYGMAG-P-SEA 20970 CZK
AXYGMAG-P-S AXYGMAG-P-M
Plasmid purification kit, Axygen® AxyPrep™ Mag
Činidla pro nukleové kyseliny Nucleic Acid Purification Kits and Reagents

AxyPrep™ Mag Plasmid Purification Kit utilises a magnetic bead technology for high throughput purification of plasmid DNA from E.coli cells. The AxyPrep™ Mag kit can also be used with fosmid and BAC vector-based constructs ranging from 5 to 150 Kb. The system consists of steps of pelleting cells, resuspension, lysis, neutralisation, DNA binding, ethanol washing and plasmid elution. The protocol is automation-compatible.

Objednat

Produktová specifikace

Popis AxyPrep™ Mag Plasmid Purification Kit
Environmentally Preferable
Aplikace
Target molecule Plasmid
Refine target molecule Plasmid
Sample type Bacteria
Method / Format Magnetic beads
Prep size Scalable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR